Sodium acetate, Corning® 3M™
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
46-033-CI
MDTC46-033-CIEA 40.1 EUR InStock
MDTC46-033-CI
Sodium acetate, Corning® 3M™
Puffer Gewebekultur-Puffer
Sodium Acetate is a common molecular biology reagent. All production lots are tested to ensure that they are free of RNAse. It is often used in buffer systems to maintain the pH of a given solution.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge