Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Glutamin, Pulver Reagenz für die Zellkultur, Corning®

Lieferant: MEDIATECH

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


61-030-RR 61-030-RO 61-030-RM
MDTC61-030-RREA 230.32 EUR InStock
MDTC61-030-RR MDTC61-030-RO MDTC61-030-RM
L(+)-Glutamin, Pulver Reagenz für die Zellkultur, Corning®
L(+)-Glutamin
L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use.

This product was tested in accordance with currently approved Mediatech, Inc. specifications and procedures. Testing procedures are maintained in compliance with the current versions of the USP and/or EP, where applicable.

Achtung! Bitte beachten: For Research or Further Manufacturing Use Only Not for use in Diagnostic Procedures.


Formel: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Siedepunkt: 355
Storage Temperature: Kühlschrank
MDL Number: MFCD00008044
CAS-Nummer: 56-85-9
Merck Index: 13,04484
Order Now
Spezifikation Testergebnisse
Appearance White crystals or crystalline powder
USP specifications
Identification Passes test
Assay 98.5% - 101.5% (calculated on dried basis)
Residue on Ignition NMT 0.1%
Chloride NMT 0.05%
Sulfate NMT 0.03%
Iron NMT 30 ppm
Heavy Metals NMT 15 ppm
Related Compounds NMT 0.5%
Specific Rotation at 20 °C +6.3 ° to 7.3 °
Loss on Drying NMT 0.3%
FCC specifications
Identification Passes test
Assay 98.5%-10_1.5% (on dried basis)
Lead NMT 5mg/kg
Specific Rotation at 20°C +6.3 ° to 7.3 °

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge