Yeast extract, powder, Corning®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
61-231-RO
MDTC61-231-ROEA 265.81 EUR InStock
MDTC61-231-RO
Yeast extract, powder, Corning®
Medien für die Mikrobiologie Trockennährböden für die Mikrobiologie
Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates obtained from autolysed yeast
This is the water soluble portion with intact B-complex vitamins and can be used as an additive for culture media.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge