Tris base buffer, powder, Corning®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
61-233-RM 61-233-RT 61-233-RR
MDTC61-233-RMEA 121 EUR InStock
MDTC61-233-RM MDTC61-233-RT MDTC61-233-RR
Tris base buffer, powder, Corning®
Puffer Elektrophorese-Puffer Laufpuffer
Tris is commonly used in buffer solutions in biochemical and molecular biological research.
This primary amine has an effective pH range of 7,0 to 9,2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge