Fructanase
Lieferant: MEGAZYME
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
E-FRMXLQ
MEGAE-FRMXLQEA 401 EUR InStock
MEGAE-FRMXLQ
Fructanase
Enzyme
This purified fructanase mixture is used for the determination of fructn (liquid), in research, biochemical enzyme assays and in vitro diagnostic analysis.
This mixture consists of exo-inulinase 2000 U/ml (on kestose, at 40 °C), endo-inulinase ∼100 U/ml (on fructan at 40 °C), α-galactosidase <0,08 U/ml (on polygalacturonic acid at 40 °C), β-Glucanase <0,04 U/ml (on β-glucan at 40 °C) and pectinase <0,07 U/ml (on pectin at 40 °C).
This enzyme is supplied in a stabilised solution containing 50% glycerol and 0.02% sodium azide.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge