Precision balance, ME4002/M
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30046451
METB30046451EA 2855 EUR InStock
METB30046451
Precision balance, ME4002/M
Waagen
State-of-the art weighing technology, solid construction and high quality materials ensure the accurate results.
  • Hygenic design
  • Easy, efficient and functional
  • Backlit LCD
  • Maximum capacity: 4,2 kg
The easy-to-use interface enables direct access to applications and also the unit is coupled with easy cleaning to ensures the fast and efficient action. The large numbers on the brilliant backlit display are easy to read in all working environments. With the built-in function, the date and time can be added to the results in accordance with ISO/GLP documentation requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge