Halogen moisture analyser, HC103
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30216101
METB30216101EA 6480 EUR InStock
METB30216101
Halogen moisture analyser, HC103
Analysatoren Feuchte-Analysatoren
The HC103 halogen moisture analyser combines proven Mettler Toledo weighing technology with fast halogen heating. Ideal for quality control and in-process control applications.
  • Intuitive touchscreen operation
  • Compact design saves space
  • Halogen heating: Accurate temperature control between 40 and 230 °C
  • High performance weighing cell for accurate moisture results
  • RS232 and USB interfaces for convenient data transfer
  • Flat stainless steel surfaces are easy to clean and robust
Users can access drying methods directly from the home screen with just 'One Click'. As the measurement is running, a real-time drying curve is displayed on the screen to provide an overview of the progress. With storage for up to 20 methods and the ability to easily transfer and upload methods via the USB port the HC103 increases productivity in the workplace.
Reports can be generated as PDF or CSV files and transferred via the RS232 or USB port to fulfil documentation and traceability requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge