Titrator Compact G20S
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30252669
METN30252669EA 0 EUR InStock
METN30252669
Titrator Compact G20S
Titratoren
The G20S extends usability with the same space saving design as the Compact titrators. Supports 150 analysis methods with 12 method functions +12 shortcuts per user, plus LabX Express software. For multiple routine titrations and users.
  • Solid Performance
  • Dependable Compact Titrators execute daily general titration analyses with multiple operators and minimum time, training and maintenance
  • Expanded Analysis Options
  • The G20S manages multiple applications with ease thanks to expanded analysis methods, method functions, shortcuts and software options
  • One Click User Interface
  • With the intuitive One Click touchscreen interface, you may customize the home screen to run your workflows quickly and easily with a single tap on the screen
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge