Dunnage rack, heavy-duty
Lieferant: Metro International
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HP52K3
METOHP52K3EA 695 EUR InStock
METOHP52K3
Dunnage rack, heavy-duty
Möbel Regale
These heavy duty tubular construction is designed to withstand impact and hold heavy loads.
  • Removable top mat lifts off for cleaning
  • Sturdy: posts and shelves mean fast, easy assembly and secure locking
  • Space Saving: platforms to store bulky items off the floor in less space than if regular shelving were used
Construction: Metroseal 3 epoxy with Microban.
Lieferumfang: Supplied with mat, cap and leveling bolt.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge