Divider and tote boxes for shelving systems
Lieferant: Metro International
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DL93080NAT DS93080NAT MTB93030W MTB93060W DL93030NAT DS93030NAT MTB93080W
METODL93080NATEA 14.7 EUR InStock
METODL93080NAT METODS93080NAT METOMTB93030W METOMTB93060W METODL93030NAT METODS93030NAT METOMTB93080W
Divider and tote boxes for shelving systems
Schalen/Schüsseln/Töpfe Einsätze für Schubladen
The tote box provide safe and efficient storage of items in bulk, or in compartmentalised fashion with the use of optional dividers and covers.
PP base resin.
  • Ideal for storage and can be transported
  • Available in various colour options
  • Injection molded for longer life and more uniform wall thickness
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge