Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30092988 30092989
METT30092988EA 7090 EUR InStock
METT30092988 METT30092989
Multi-parameter meters, bench, SevenExcellence™ S479 / S975-uMix
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
Modular triple-channel instruments with touch screen operation, which can easily handle the most complex applications and meet stringent requirements in regulated markets.
  • Allows for measurements of 1, 2 or 3 parameters simultaneously, either in the same or different samples
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • uPlace™ electrode holder (included) can be operated with one hand
  • Large 7 inch colour display and intuitive menu guidance in 10 languages
  • EasyDirect™ pH PC software for transferring real time measurement data and archiving the results
  • GLP support functions: PIN protection, GLP printout format with all GLP relevant information and monitoring of measurement limits
S479 allows users to measure pH/mV, rel. mV/ORP, conductivity (TDS, salinity, resisitivity, conductivity ash) as well as dissolved oxygen (using digital or polarographic technology).
S975 allows users to measure pH/mV, rel. mV/ORP, conductivity (TDS, salinity, resisitivity, conductivity ash), dissolved oxygen (using digital or polarographic technology), and the concentration and activity of ions in solution.
SevenExcellence meters fit three modules (expansion units) in any composition and order. They offer the highest possible flexibility as users can expand the instrument at any time, adding new measurement parameters.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge