Ion selective electrodes and half-cells
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
51340263 51107672 51107673 51107674 51107675 51107676 51107678 51107680 51107681 51107686 51107687 51107688 51107689 51107690 51107692 51107694 51107695 51107870 51107871 51107873 51107874 51089935
662-2803EA 490 EUR InStock
662-2803 METT51107672 METT51107673 METT51107674 METT51107675 METT51107676 METT51107678 METT51107680 METT51107681 METT51107686 METT51107687 METT51107688 METT51107689 METT51107690 METT51107692 METT51107694 METT51107695 METT51107870 METT51107871 METT51107873 METT51107874 METT51089935
Ion selective electrodes and half-cells
Elektroden ISE-Elektroden
These ion-selective half-cells (exception: Sodium-sensitive electrode) consist of two elements: a universal shaft and an ion-specific membrane module.
  • Available as half-cells (require external reference electrode) or as electrodes with built-in reference
  • Each ion-selective electrode (ISE) and each membrane module is tested wet-chemically
  • Each ISE is issued with its own serial number and quality certificate
  • Electrodes are filled with electrolyte and ready for use
  • Membrane kit comes with an identification ring (ID ring) and an adapter sleeve which will fit any METTLER TOLEDO titration stand
This module may be exchanged allowing users to measure the ion of their choice. They just need to order the membrane kit specific for that ion, mount the new module onto the shaft of the ISE half-cell, and they have a new ISE.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge