Parker Hannifin Push-to-Connect Check Valves, Acetal
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01379-89 01379-93
MFLX01379-89EA 22.5 EUR InStock
MFLX01379-89 MFLX01379-93
Parker Hannifin Push-to-Connect Check Valves, Acetal
Ventile Rückschlagventile
Create a secure connection with your check valve.
  • Constructed of durable acetal plastic
Use these push-to-connect check valves for rigid and semirigid plastic tubing. Acetal body with EPDM O-rings. Temperature range of 34 °F (1.1 °C) to 150 °F (65.6 °C).
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge