Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06040-45 06040-46 06040-53
MFLX06040-45EA 110 EUR InStock
MFLX06040-45 MFLX06040-46 MFLX06040-53
Bürkle Hand-Operated Pump Dispensers for Commercial Storage Drums
Pumpen Fass/Transferpumpen
Dispense liquids safely and quickly from most storage containers.
  • Telescopic immersion tube adapts itself to a variety of containers
  • Different materials and styles suit a wide range of applications
These hand-operated pumps feature a universal soft PVC bung consisting of a conical center and two conical sleeves allowing for an airtight seal between the pump and most liquid storage container openings. The telescopic immersion tube adapts itself to the container height on insertion, ensuring good removal of residual liquid. Various tube diameters are available to meet your individual liquid transfer needs. Choose from polypropylene (PP), PVDF, or 304 stainless steel (SS) wetted materials and dispensing by spout or hose.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge