Hand-Operated Drum Pumps, HDPE
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06425-05
MFLX06425-05EA 307 EUR InStock
MFLX06425-05
Hand-Operated Drum Pumps, HDPE
Pumpen Fass/Transferpumpen
Ideal for most acid/base or ionic salt applications.
  • Pump acids, alcohol, and alkalies from carboys, drums, or tanks
  • Self-cleaning ball valve ensures free flow and long pump life
This plastic hand-operated drum pump is ideal for pumping acids, alcohols, and alkalis. Constructed of all HDPE parts, this pump offers 6 strokes/gallon (2 strokes/liter). Order optional LDPE adapter to securely mount pump on drums. Adapter 06425-05 sold separately.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge