Hand-operated detergent, waxes, alkalis and acid transfer pump
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06433-31
MFLX06433-31EA 184 EUR InStock
MFLX06433-31
Hand-operated detergent, waxes, alkalis and acid transfer pump
Pumpen Fass/Transferpumpen
Ideal for the janitorial market for transfering detergents.
  • Tested for transferring HCl in concentrations up to 30% at 100 °F (38 °C)
  • Self-priming, will not siphon
This hand-operated drum pump is ideal for pumping detergents, waxes, alkalis and certain acids. Constructed of solvent-welded PVC body and shaft, polyethylene (PE) hose, Viton® valves and seals, this pump offers 5 strokes/gallon.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge