Gast® Direct-Drive Hub-Mount Air Motors
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
07055-70
MFLX07055-70EA 395 EUR InStock
MFLX07055-70
Gast® Direct-Drive Hub-Mount Air Motors
Pumpen Motoren
Four-vane rotary design is almost maintenance free.
  • Nonsparking, nonelectrical operation—safe and reliable
These Gast® Direct-Drive Hub-Mount Air Motors feature a 1/2 hp reversible motor.
Lieferumfang: Includes motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and filters separately.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge