Masterflex® Single Use Tubing Pinch Clamps, Avantor®
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MFLX19999-96EA 36.8 EUR InStock
MFLX19999-96
Masterflex® Single Use Tubing Pinch Clamps, Avantor®
Klemmen Schlauchklemmen und Sanitärklemmen
Clamps are double bagged and ready for use in your cleanroom environment.
  • Sturdy glass reinforced nylon design
  • Validated to USP Class VI
  • Rated to 100 PSI (6,5 bar)
  • Feature anti-tamper zip-tie lock out
  • Sterilisable via autoclave or gamma irradiation (25 to 43 kG)
  • Features removable sled for locking clamp in place
  • Injection molded and assembled in ISO cleanroom
These clamps are manufactured, assembled, and packaged within an ISO cleanroom.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge