Cadence Luer Fittings, 316 Stainless Steel
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
41507-20 41507-21 41507-22 41507-23 41507-24 41507-25 41507-26 41507-27 41507-28 41507-29 41507-30 41507-31 41507-32 41507-33 41507-34 41507-44 41507-45 41507-46 41507-48 41507-89 41507-90
MFLX41507-20EA 56.6 EUR InStock
MFLX41507-20 MFLX41507-21 MFLX41507-22 MFLX41507-23 MFLX41507-24 MFLX41507-25 MFLX41507-26 MFLX41507-27 MFLX41507-28 MFLX41507-29 MFLX41507-30 MFLX41507-31 MFLX41507-32 MFLX41507-33 MFLX41507-34 MFLX41507-44 MFLX41507-45 MFLX41507-46 MFLX41507-48 MFLX41507-89 MFLX41507-90
Cadence Luer Fittings, 316 Stainless Steel
Fittings und Verbinder
Exceptional durability.
  • Easy to disassemble for cleaning
  • Ideal for harsh chemical environments
  • Autoclavable
Constructed of durable 316 stainless steel, these Luer Adapter Fittings are ideal for a wide range of applications.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge