Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MFLX45518-82BLKEA 408 EUR InStock
MFLX45518-82BLK MFLX45518-88BLK
Value Plastics® Adapter Fittings, Male Luer to Threaded, Straight
Fittings und Verbinder
Easily transition between luer fittings and UNF or NPT threads.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge