Hand-Operated FDA Compliant Drum Pumps
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70607-20
MFLX70607-20EA 106 EUR InStock
MFLX70607-20
Hand-Operated FDA Compliant Drum Pumps
Pumpen Fass/Transferpumpen
Ideal for most acid/base or ionic salt applications.
  • Materials are FDA compliant for indirect food contact
  • Available with outlet hose (70607-20) or spout (70607-22)
These plastic hand-operated drum pumps are constructed with materials that conform under FDA CFR 21 regulations for indirect food contact. Pumps features 26 strokes/gallon (7 strokes/liter) with 3/4” NPT(M) container adapter. Both pumps are constructed with PVC barrel, Viton® valves and seals, and LDPE tube. Choose between models with a PVC hose outlet or LDPE spout outlet.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge