ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
74020-00
MFLX74020-00EA 3320 EUR InStock
MFLX74020-00
ITT Jabsco High-Purity Air-Operated Double-Diaphragm Pumps
Pumpen Druckluftpumpen
Unique air valve ensures a positive shift every time – pump will not stall.
  • All diaphragms, liquid chambers, and manifolds are constructed
  • of 100% virgin PTFE for long life and high purity
  • Unique valve requires no lubrication or prepacking.
  • Easily access air valve without disassembling the pump
Pumps are manufactured under class 1000 clean room conditions to guarantee purity.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge