White Epoxy Motor Sanitary Centrifugal Pumps
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
76710-10
MFLX76710-10EA 3420 EUR InStock
MFLX76710-10
White Epoxy Motor Sanitary Centrifugal Pumps
Pumpen Kreiselpumpen
Clean-in-place (CIP) 3A sanitary transfer pump.
  • Washdown motors available in white epoxy
  • Stainless steel adapters are standard
  • Ideal for RO water and pharmaceutical manufacturing
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
Lieferumfang: Includes conduit box for hardwiring.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge