Direct Coupled Brass-Bodied Rotary Vane Pumps
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
79410-05 79410-10 79410-15
MFLX79410-05EA 604 EUR InStock
MFLX79410-05 MFLX79410-10 MFLX79410-15
Direct Coupled Brass-Bodied Rotary Vane Pumps
Pumpen Flexible/Rotations-/Roller-Flügelradpumpen
Ideal for high pressure applications—up to 240 psi.
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge