Masterflex® Solenoid-Operated Pinch Valves, Avantor®
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
98302-00 98302-02 98302-06 98302-10 98302-12 98302-16 98302-20 98302-22 98302-26 98302-32 98302-36 98302-40 98302-42 98302-46 98302-48 98302-50 98302-54 98302-30
MFLX98302-00EA 104 EUR InStock
MFLX98302-00 MFLX98302-02 MFLX98302-06 MFLX98302-10 MFLX98302-12 MFLX98302-16 MFLX98302-20 MFLX98302-22 MFLX98302-26 MFLX98302-32 MFLX98302-36 MFLX98302-40 MFLX98302-42 MFLX98302-46 MFLX98302-48 MFLX98302-50 MFLX98302-54 MFLX98302-30
Masterflex® Solenoid-Operated Pinch Valves, Avantor®
Ventile Solenoidventile
No contamination of valve.
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge