Cleanroom autoclave indicator tape
Lieferant: MICRONOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AUTOPC-1 AUTOPC-34 AUTOPC-15 AUTOPC-12 AUTOPC-2 AUTOPC-3
MICRAUTOPC-1EA 38.9 EUR InStock
MICRAUTOPC-1 MICRAUTOPC-34 MICRAUTOPC-15 MICRAUTOPC-12 MICRAUTOPC-2 MICRAUTOPC-3
Cleanroom autoclave indicator tape
Sterilisationsindikatoren
Saturated crepe paper/synthetic poly isoprene adhesive in plastic core. It is a proven method of positively confirming a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. Green indicator lines change to black when proper levels of steam pressure and temperature have been achieved. After use, the tape removes quickly and completely, leaving minimal residue or staining.
  • Lead and latex-free
  • RoHS compliant
  • Uniquely printed lines cannot be rubbed off and will not contaminate instruments
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge