Bucket Liners
Lieferant: MICRONOVA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BKL993521IRB
MICRBKL993521IRBEA 0 EUR InStock
MICRBKL993521IRB
Bucket Liners
Beutel Entsorgungsbeutel
The unique LDPE/nylon laminate is a superb barrier material for lining totes to transport raw materials or to line buckets containing disinfectants or other cleaning solutions.
  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts or raw material transfer
  • Thickness of 3 mil
  • Will hold up to autoclaving with liquid contents
  • Available sterile (irradiated) or non sterile
Non sterile liners are bulk packed. Select sterile liners are available bulk packed.
Fits: B-99 series buckets.
Verpackung: Flat packed, either individually double-bagged or in bulk packs. Packed in a case of 24 or case of 72 for bulk pack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge