Mobile Phase Filters
Lieferant: MICROSOLV
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
58846-05
MICS58846-05EA 49 EUR InStock
MICS58846-05
Mobile Phase Filters
Filter Flüssigkeits-Prozessfilter
Biocompatible, last drop, PEEK and polypropylene mobile phase filters. PTFE filter element and will provide very high filter capacity and throughput while using a maximum amount of solvent. The unique design allows for this flat filter to vacuum up to the last drop of liquid in the reservoir bottle.
  • Tubing connector uses the fitting and an ETFE ferrule to hold non-flanged low pressure tubing in the filter without leaking
  • The 5 µm pore size element will filter undissolved particles from your solvents that are 5 µm or larger preventing them from damaging your HPLC pumps, columns or instrument
  • ARE-applied research brand one each
The advantage of a white filter element is that they make it easy to quickly see particles that have been trapped by the filter and if the element needs to be replaced to maintain proper flow of your solvents. These filter elements are soft and easy to remove and replace.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge