Autosampler vials, 3,8 ml
Lieferant: MICROSOLV
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9534S-0CV
MICS9534S-0CVEA 318 EUR InStock
MICS9534S-0CV
Autosampler vials, 3,8 ml
Vials
These vials made of TYPE 1 borosilicate glass. The unique center drain offers maximum recovery of compounds placed in these vials for storage or sampling.
  • Sturdy with a 'trumpet' bottom
  • Ultra low dead volume for better recovery with center drain
  • Suitable for use with Waters autosamplers
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge