Amicon™ XM Ultrafiltration Membranes Dics Filters
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
14112AM
MILF14112AMEA 263 EUR InStock
MILF14112AM
Amicon™ XM Ultrafiltration Membranes Dics Filters
Filter Membranfilter
Ultrafiltration membranes - Amicon™ XM polyacrylonitrile/PVC co-polymer filters. High-recovery membranes for stirred cells.
  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge