Glass fiber filters with binder resin
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AP2504700 AP1502500 AP1507500 AP1509000 AP1512450 AP1514250 AP2001300 AP2004200 AP2005500 AP2007500 AP2009000 AP2012450 AP2014250 AP2501000 AP2502200 AP2502500 AP2509000 AP2512450 AP2004700 AP2504200 AP1504200 AP2507500 AP2514250 AP2501300 AP1504700 AP2002500
MILFAP2504700EA 75.5 EUR InStock
MILFAP2504700 MILFAP1502500 MILFAP1507500 MILFAP1509000 MILFAP1512450 MILFAP1514250 MILFAP2001300 MILFAP2004200 MILFAP2005500 MILFAP2007500 MILFAP2009000 MILFAP2012450 MILFAP2014250 MILFAP2501000 MILFAP2502200 MILFAP2502500 MILFAP2509000 MILFAP2512450 512-0242 MILFAP2504200 MILFAP1504200 MILFAP2507500 MILFAP2514250 MILFAP2501300 MILFAP1504700 MILFAP2002500
Glass fiber filters with binder resin
Filter Glasfaserfilter
Millipore filters with binder resin have superior wet strength and are excellent for qualitative analysis and prefiltration, especially for heavily contaminated liquids. They are also widely used for clarification of aqueous solutions.
Type AP15: Low dirt-holding capacity. Recommended prefilter for 0,2 to 0,6 µm filters.
Type AP20: Low retention and high dirt-holding capacity than AP15 filters. Recommended prefilter for 0,8 to 8,0 µm filters. Use upstream to protect type AP15 filters.
Type AP25: Added thickness yields same retention and greater dirt-holding capacity as AP20. Recommended prefilter for 0,9 to 8 µm filters, especially for proteinaceous and heavily contaminated liquids. Use upstream to protect type AP15 filters.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge