Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Membrane filters, LCR

Lieferant: Merck

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

FHLC01300 FHLC02500 FHLC04700
MILFFHLC01300EA 332 EUR InStock
MILFFHLC01300 MILFFHLC02500 512-0247
Membrane filters, LCR
Filter Membranfilter
These PTFE white hydrophilic membrane compatible with all commonly used HPLC solvents. The membrane undergoes a special treatment process to remove any residual extractables, to provide clearer analysis results. They are widely used for HPLC mobile phase filtration, clarifying acids, bases and dilute protein solutions and isolation of RNA.

  • Hydrophilic membrane can be used to filter aqueous fluids without prior wetting

Filter surface: Plain; Thickness: 140 µm

Operating temperature: 130 °C

Gravimetric extractables: 0,5%

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge