Immobilon®-FL PVDF Transfer Membranes
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
IPFL07810
MILFIPFL07810EA 120 EUR InStock
MILFIPFL07810
Immobilon®-FL PVDF Transfer Membranes
Transfer-Membranen
The Immobilon®-FL transfer membrane is a polyvinylidene fluoride (PVDF) microporous membrane for binding proteins transferred from a variety of gel matrices.
The Immobilon®-FL membrane has a nominal pore size of 0,45 μm and is recommended for blotting proteins with molecular weights greater than 20 kDa. The membrane exhibits very low autofluorescence across a wide range of excitation/emission wavelengths. This property makes it ideal for blotting applications involving fluorescence-based immunodetection.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge