Aerosol analysis monitors
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AP1002500 MAWP025A0 MAWP025AC
MILFAP1002500EA 59.1 EUR InStock
MILFAP1002500 MILFMAWP025A0 MILFMAWP025AC
Aerosol analysis monitors
Filter Kassettenfilter
The PS monitor is used for sample collection and particle analysis. The PP monitor is used for microscopic particle analysis and sample collection for HPLC and UV detection.
Membrane: mixed cellulose esters with cellulose support pad; housing: PS or carbon filled PP.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge