Filter holders, Hydrosol™
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
XX2004720
MILFXX2004720EA 1710 EUR InStock
MILFXX2004720
Filter holders, Hydrosol™
Filterhalter
This filter holder is suitable for particulate and biological contamination analysis via vacuum filtration.
  • Not autoclavable with filter in place
Lieferumfang: Includes grounding set for use with flammable liquids.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge