In-line filter holders, stainless steel
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
XX4002500 XX4504700 XX4502500 XX4404700
MILFXX4002500EA 463 EUR InStock
MILFXX4002500 MILFXX4504700 MILFXX4502500 MILFXX4404700
In-line filter holders, stainless steel
Filterhalter
Stainless steel in-line filter holders for filtering liquids or gases through 25 mm or 47 mm disc filters at pressures up to 700 bar.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge