Polysep II Opticap XL T-line capsule filters - 1µm/ 0,2µm - Length 20 inch - Borosilicate glass microfiber and mixed esters of cellulose Filter Media - 0,92m² - 16 mm (5/8 ) Hose barb Inlet and Outlet - 1 per package 1 * 1 ST
Lieferant: MILLIPORE
Polysep II Opticap XL T-line capsule filters - 1µm/ 0,2µm - Length 20 inch - Borosilicate glass microfiber and mixed esters of cellulose Filter Media - 0,92m² - 16 mm (5/8 ) Hose barb Inlet and Outlet - 1 per package 1 * 1 ST
New Product
PRODUKT
Erweitert / Kompakt
Polysep II Opticap XL T-line capsule filters - 1µm/ 0,2µm - Length 20 inch - Borosilicate glass microfiber and mixed esters of cellulose Filter Media - 0,92m² - 16 mm (5/8 ) Hose barb Inlet and Outlet - 1 per package 1 * 1 ST MILXKGW3A2THH1 1 * 1 ST
New Product
Hersteller-Artikelnummer:: KGW3A2THH1
Lokale Artikelnummer:: MILXKGW3A2THH1
Barcode erstellen Barcode Label Format: 
PDF Katalogseiten erstellen
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder