Milliflex Oasis® Vacuum Pump
Lieferant: Merck
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MMSYSTMM1
MILXMMSYSTMM1EA 13000 EUR InStock
MILXMMSYSTMM1
Milliflex Oasis® Vacuum Pump
Pumpen Vakuum-Membranpumpen
The Milliflex Oasis® vacuum pump made of acrylonitrile styrene acrylate and polycarbonate.
  • Quick connections for power and drain tubing
  • No calibration needed
  • Low height - 70 mm (2,8")
  • Check valve situated in the removable filtration head prevents potential contamination
  • Accessible key parts of the head for easy cleaning
  • No autoclave is required for hardware decontamination
The Milliflex Oasis® vacuum pump consists of a controlled vacuum source and filtration support. The system is design for biosafety cabinets/laminar flow hoods, with its small footprint, low weight and easy to decontaminate surfaces. This uniquely designed pump reduces the risk of cross-contamination and false results. Both ergonomics and scalability generate high throughput. No autoclave is required for hardware decontamination, only monthly sanitisation, to dramatically reduce the workload in the lab.
Scalability: Connect up to 3 pumps and start up to 6 simultaneous filtrations with a single push of the button.
It offers complete solution for bioburden testing, including services, for a secured workflow with the highest throughput capability: All-in-one filtration equipment, ready-to-use sterilized filtration units, pharmacopeia compliant culture media.
Apllications: It has used in water testing/bioburden testing, cosmetics and pharmaceutical industries.
Order Now
PRODUKT
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge