Calcium assay kits, FLIPR® Calcium 4 assay kit
Lieferant: MOLECULAR DEVICES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R8142 R8143 R8141
MLDVR8142EA 910 EUR InStock
MLDVR8142 MLDVR8143 MLDVR8141
Calcium assay kits, FLIPR® Calcium 4 assay kit
Tests Zelltests
The FLIPR® Calcium 4 Assay Kit provides a fast, simple and reliable fluorescence-based assay for detecting changes in intracellular calcium.
  • Maximises fluorescent signal by reducing cell wash artifacts
  • Reduced well-to-well variability, improving assay quality (Z' factor) and reliability (CV %) of high-throughput screens
  • Mix-and-read protocol accelerates assay workflow and increases throughput
  • Superior signal-to-noise ratio facilitates confirmation of endogenous or transiently transfected receptor activity during assay development
  • Pre-optimised and validated protocols for the FLIPR® Tetra and FlexStation® Systems ensure navigation to both routine and unconventional cell lines and targets
FLIPR® Calcium 4 Assay Kit contains a proprietary masking dye (Component A) with higher light extinction in the extracellular solution to significantly reduce background and increase signal window. It is more inert and reduces compound interference. Successful targets include calcium channels, chemokine receptors, and transiently transfected receptors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge