IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Lieferant: MOLECULAR DEVICES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R8160
MLDVR8160EA 0 EUR InStock
MLDVR8160
IMAP kinase, phosphatase, and phosphodiesterase assays, molecular devices
Tests Enzymtests
IMAP® technology provides a homogeneous assay applicable to a wide variety of kinases without regard for substrate peptide sequence.
  • Complete assay system
  • Robust fluorescence signal
  • Homogeneous
  • Available in both FP and TR-FRET detection modes
IMAP provides a complete assay system for screening kinases, phosphatases, and phosphodiesterases
The assay is a simple "mix-and-read" procedure that allows accurate determination of kinase, phosphatase, and phosphodiesterase (PDE) activity.
IMAP Binding Reagent and Binding Buffers for 8000 data points in a standard 384 well format.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge