Calcium assay kits, Fura-2 QBT™ Calcium kit
Lieferant: MOLECULAR DEVICES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R8197 R8198
MLDVR8197EA 562 EUR InStock
MLDVR8197 MLDVR8198
Calcium assay kits, Fura-2 QBT™ Calcium kit
Tests Zelltests
The Fura-2 QBT™ Calcium Kit eliminates the cause of data variability and reduces the number of steps compared to conventional wash protocols using Fura-2.
  • Largest Fura-2 signal window available
  • Eliminate wash artifacts and increase throughput with homogeneous assay
  • Minimise the impact of uneven dye loading and leakage on results
  • Interrogate low density, weakly or non adherent cells using no-wash protocol
  • Assay on FlexStation® 3 and FLIPR® Tetra Systems
  • Reference thousands of citations where Fura-2 has been used for cell lines and target
Lieferumfang: For researchers concerned about assay artifacts, the Fura-2 QBT™ Calcium Kit utilises a simple, mix-and-read format that employs a proprietary masking technology with the industry-standard Fura-2 ratiometric calcium indicator to accurately measure calcium mobilisation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge