Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
Lieferant: MOLECULAR DEVICES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R8361
MLDVR8361EA 369 EUR InStock
MLDVR8361
Reporter gene assays, SpectraMax® DuoLuc™ reporter assay kit
Tests Zelltests
SpectraMax® DuoLuc™ reporter assay kit offers sensitive detection of dual luciferase reporter gene assays, which are important tools for monitoring gene expression and other cellular events.
  • Enables highly sensitive quantitation of both firefly and Renilla luciferases in mammalian cells
  • Flash-type luminescence with excellent sensitivity
  • Expression measurement in as few as ten cells per well
  • Improved accuracy of results by normalising reporter expression to a control
  • Optimised for SpectraMax® i3x, iD3, or L microplate readers with injectors
  • Simplified data acquisition and analysis with preconfigured protocol in SoftMax® Pro Software
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge