UHMW PE Film Tape, 5421
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5421101
MMMA5421101EA 624 EUR InStock
MMMA5421101
UHMW PE Film Tape, 5421
Klebebänder
3M™ UHMW PE Film Tape 5421 protects surfaces from abrasion. This tape has a thickness 0,01 mm that offers excellent abrasion resistance, making it ideal for many tough impact wear or sliding applications. It creates a 'slip plane' effect to allow contact without wear. This is a thinner version of 3M™ UHMW-PE Film Tape 5423.
  • Excellent abrasion resistance
  • Achieve low coefficient of friction for ‘slip plane’ effect between surfaces
  • Reduce squeaks, rattles, and other noises that occur with movement
  • Use in applications where lubrication is not possible because of contamination
  • Liner makes it ideal for fabricating or die-cutting into various lengths and shapes
Use 3M™ UHMW PE Film Tape 5421 for tough impact wear or sliding applications. This general-purpose polyethylene tape has outstanding abrasion resistance to help protect surfaces from wear. It has a low coefficient of friction for ‘slip plane’ effect between surfaces and can help reduce squeaks, rattles, and other noises that occur with movement. Use it in applications where lubrication is not possible because of contamination. The tape’s liner makes it ideal for fabricating or die cutting into various lengths and shapes.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge