Fit Test Adapters
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
601
MMMA601EA 223 EUR InStock
MMMA601
Fit Test Adapters
Atemschutzmasken
For an alternative method to sharing a probed fit testing respirator, use the 3M™ quantitative fit test adapter. The adaptor kits allow users to fit test with a personal 3M™ Respirator, rather than using a shared probed fit test respirator.
  • Adapter allows fit testing with an individual's personal 3M™ respirator
  • Alternative to sharing a probed fit testing respirator
  • Quick and easy to fit
The 3M™ Quantitative fit test adapters are designed for use with 3M™ half and full-face respirators with bayonet-style filter attachments.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge