Transferklebebänder, ATG 924, Scotch®
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
924EU-12MM 924EU-19MM
MMMA924EU-12MMEA 916 EUR InStock
MMMA924EU-12MM MMMA924EU-19MM
Transferklebebänder, ATG 924, Scotch®
Klebebänder
Apply Scotch® ATG adhesive transfer tape 924 for various substrate or material combinations. This transfer film tape is an acrylic, reverse wound, pressure sensitive adhesive on a densified kraft paper liner. Use with Scotch® ATG adhesive applicators.
  • Provides a clean, permanent bond to lightweight materials
  • Get high adhesion, good solvent resistance, and excellent ultraviolet (UV) resistance
  • Bonds on contact, and works on a variety of substrate or material combinations
  • Leaves no mess, no residue, and no odor
  • Length: 33 m
  • Transfer tape back material
This transfer film tape is a great general-purpose adhesive tape, offering high adhesion, good solvent resistance, and excellent ultraviolet resistance. It bonds on contact when delivered through the ATG system. A controlled application of the adhesive is delivered while the liner rewinds into the ATG applicator. The ATG tape system is a cost effective replacement to rivets, staples, screws, bolts, or liquid adhesives, and is safer than hot melt adhesives. It’s versatile, convenient, quick, and easy to use.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge