Doppelseitiges Schaumklebeband 9515W
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9515W12
MMMA9515W12EA 9880 EUR InStock
MMMA9515W12
Doppelseitiges Schaumklebeband 9515W
Klebebänder
3M™ Double coated foam tape 9515W is a white, 1,5 mm thick, double coated polyethylene foam tape with acrylic adhesive.
  • Closed-cell double coated polyethylene foam with acrylic adhesive
  • Mount and join a wide range of materials
  • Suitable for different surfaces
The acrylic adhesive provides good initial tack and adhesion to a wide variety of surfaces. Intended applications include assembly of displays (POS), lightweight emblems, name plates, etc.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge