Hauben für motorbetriebene Atemschutzgeräte, Versaflo™
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M448
MMMAM448EA 297 EUR InStock
MMMAM448
Hauben für motorbetriebene Atemschutzgeräte, Versaflo™
Atemschutzmasken Atemschutz-Systeme
The Versaflo™ Heavy Duty Shroud M-448 is a high durability shroud for 3M™ Versaflo™ M-400 helmets. A 3M™ Versaflo™ Inner Collar (M-444) must be zipped into the outer shroud before use.
  • Provides heavy-duty, durable protection
  • Designed for use with 3M™ Versaflo™ M-400 Helmets
Get added protection while wearing 3M™ Versaflo™ M-400 Helmets with the Versaflo™ Heavy Duty Shroud, M-448. A 3M™ Versaflo™ Inner Collar (M-444) must be zipped into the outer shroud before use.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge