PELTOR™ LiteCom Plus Headset
Lieferant: 3M
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MT73H7A4410EU
MMMAMT73H7A4410EUEA 1500 EUR InStock
MMMAMT73H7A4410EU
PELTOR™ LiteCom Plus Headset
Kapselgehörschützer Kapselgehörschützer mit Radio
3M™ PELTOR™ LiteCom Plus Headset is our robust hearing protectors with a built-in analogue two-way communication radio, noise cancelling dynamic microphone, Lithium-Ion battery and level dependent functionality for ambient listening. Available in headband and helmet mounted models.
  • Noise cancelling dynamic microphone provides voice-operated transmission for hands free use
  • External connection jack allows connection of a mobile device or another two-way radio
  • Lithium-Ion battery
  • Auto power off and clear battery status indication
  • Available in headband and helmet mounted models
  • Available in 16 channel PMR 446 MHz version with 121 sub-channels
3M™ PELTOR™ LiteCom Plus Headset feature a built-in analogue two-way radio, available in PMR version with 16 channels and 121 sub-channels - to help you communicate even more efficiently in noisy environments. The noise cancelling dynamic microphone provides voice-operated transmission (VOX) for hands free use, and an external jack allows you to connect a mobile device or another two-way radio. Sound levels constantly change in demanding situations.
When the noise quiets down, you can turn up the environmental listening function to improve situational awareness or speak with co-workers who are close by, and if the hazardous noise returns the level dependent technology will kick in to help protect your hearing again. Litecom Plus Headsets have a clear battery status indicator and an auto power off function to save energy.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge