Casting tape, Scotchcast™ Plus
Lieferant: 3M MEDICAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M82002A
MMMEM82002AEA 564 EUR InStock
MMMEM82002A
Casting tape, Scotchcast™ Plus
Erste-Hilfe-Ausstattung Verbandskästen-Bestandteile
This casting tape consists of a knitted fiberglass fabric impregnated with polyurethane resin. Exposure to moisture or water initiates a chemical reaction which causes the tape to become rigid.
  • Finished casts produced from this casting tape are lightweight, strong, radiolucent, and porous
  • Scotchcast™ Plus casting tape contains a lubricating agent in the resin to provide a slippery feel when exposed to water
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge