Water ATP free swabs, Clean-Trace™
Lieferant: 3M Food Safety
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
AQF100
MMMPAQF100EA 577 EUR InStock
MMMPAQF100
Water ATP free swabs, Clean-Trace™
Tupfer Tupfer für die Probenahme
Water hygiene test to measure free ATP in a sample.
  • Innovatively designed for water sample collection uniformity
  • Detects the presence of biological contamination in Clean in Place (CIP) rinse water or other process waters
  • Self-contained 30-second test for use with CIP rinse water or other process waters
  • Allows you to understand the hygiene status of your CIP system in seconds
  • Helps reduce risk of cross contamination due to poor cleaning
  • Allows optimisation of Biocide programs
Real-time ATP water hygiene test which gives an objective indication of cleanliness in CIP water samples in seconds. A dedicated ATP water test that is designed to prevent sampling inaccuracy inherent in collecting a water sample with a swab. Its overall simplicity makes it a powerful addition to your hygiene program. Results measured in the 3M™ Clean-Trace™ NG Luminometer and provided in Relative Light units (RLU).
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge